missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 96-Well Microtiter™ Microplates

Use these 96-well microplates for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.

Brand:  Thermo Scientific™ 2205

Code : NEW

Additional Details : Weight : 3.30000kg

 View more versions of this product

Product Code. 15973620

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.

Features:

  • Use for high-throughput screening and research
  • Clear plates for colorimetric applications
  • Standard footprint, 128 × 86mm
  • Available as both polystyrene square wells and vinyl square wells
  • Vinyl Microtiter microplates are used for complement fixation, agglutination and blood typing
  • Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing and blood typing

Ordering Information:

Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.

Specifications

Specifications

Hydrophobic
Without Lid
50 Cases, 10 Pk.
Clear
Polystyrene
96
Non-treated
50
50/Pk.
Standard
8 x 12
Microtiter
Microplate
400 μL
Non-sterile
Round Bottom (U-well)
10
U96
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers