Learn More
Thermo Scientific™ 96-Well Microtiter™ Microplates
Use these 96-well microplates for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Brand: Thermo Scientific™ 2101
Code : 04
Additional Details : Weight : 1.81800kg
Description
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
- For high-throughput screening and research
- Clear plates for colorimetric applications
- Standard footprint, 128 × 86mm
- Available as both polystyrene square wells and vinyl square wells
- Vinyl Microtiter microplates are used for complement fixation, agglutination, and blood typing
- Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing, and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
Specifications
Alphanumeric | |
Without Lid | |
25 Pk., 100 Cases | |
Clear | |
Vinyl | |
96 | |
Non-treated | |
100 |
Standard | |
8 x 12 | |
Microtiter | |
Microplate | |
400 μL | |
Non-sterile | |
Round Bottom (U-well) | |
25 |