missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Non-treated 96-well Microplates
Manufactured without surface treatment to create low biomolecular binding for sensitive biochemical screening applications. Greiner Bio One™ Non-treated 96-Well Assay Microplates are available in polystyrene or polypropylene with a range of well designs.
£98.79 - £116.60
Specifications
Barcode | Alphanumeric |
---|---|
Certifications/Compliance | DIN ISO 9001 |
Format | Standard |
Lid | Without Lid |
Array | 8 x 12 |
Product Code | Brand | Total Volume (Metric) Well | Material | Well Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Total Volume (Metric) Well | Material | Well Shape | Price | Quantity & Availability | |||
10008800
|
Greiner Bio-One
650101 |
323 μL | Polystyrene | Round Bottom (U-well) |
£98.79
Pack of 100 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10252711
|
Greiner Bio-One
655101 |
382 μL | Polystyrene | Flat Bottom (F-well) |
£116.60
Pack of 100 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||
Description
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- Free of detectable endotoxins (0.003 EU/mL) and regularly tested using an FDA-approved kinetic turbidimetric LAL-test (Limulus Amoebocyte Assay)
- Manufactured from raw materials tested for leachables
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Models with chimney design allow each well to stand on its owndiminishing the risk of sample carryover and cross contamination
Specifications
Alphanumeric | |
Standard | |
8 x 12 | |
96 | |
Non-treated | |
100/Cs. |
DIN ISO 9001 | |
Without Lid | |
Clear | |
Non-sterile | |
Yes |