Learn More
Greiner Bio-One 96-Well Microplate CELLSTAR™, Cell Culture-Treated
Product Code. 32043092
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
Unit Size:
Pack of 32
Product Code. Well Shape Unit Size
32043092 Flat Bottom (F-well) Pack of 32
1 options
Product Code. 32043092 Supplier Greiner Bio-One Supplier No. 655090GBC
  • ONLINE EXCLUSIVE PRICE
    £254.25
    £234.24 / Pack of 32
    Save £20.01
    7% Off
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for cell culture and microscopy using fluorescence or luminescence detection. Optimized for low to medium magnification.
  • For barcoded versions: Unique Code 128 barcode on all four sides of the microplate, consisting of a plate type-specific 3-letter prefix followed by a unique 7-digit serial number
  • ANSI-compliant footprint for compatibility with automated systems
  • Physical surface treatment for improved cell adhesion
  • Enhanced cell attachment and higher proliferation rates to improve and accelerate cell expansion
  • Free of detectable DNase, RNase, and human DNA
  • Non-pyrogenic
  • Non-cytotoxic
  • High optical clarity for optimal microscopic examination
  • Stackable design for efficient storage and handling
  • Chimney well design allows each individual well to stand on its own, minimizing the risk of sample carryover and cross-contamination
  • Manufactured under DIN ISO 9001 guidelines
  • Supplied in a pack of 32
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Format Standard
Array 8 x 12
Color Black, Clear Bottom
Product Type Pre-barcoded Plate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Tissue Culture
Well Shape Flat Bottom (F-well)