missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Half Area ELISA Microplates

Click to view available options
Sterility:
Non-sterile
Sterile
Surface Treatment:
Microlon 600 High Binding
Microlon Medium Binding
Non-treated
Unit Size:
Pack of 40
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Crystal clear polystyrene
- Alphanumeric well coding
- Suitable for transmission measurements
- Well-to-well spacing: 9mm
- Specifically tre
Certifications
DIN ISO 9001
Specifications
Specifications
| Format | Half-Area |
| Array | 8 x 12 |
| Binding Type | Medium Binding |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Microlon Medium Binding |
| Well Shape | Flat Bottom (F-well) |
| No. per Pack | 10 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction