Learn More
Greiner Bio-One 96-well Half Area ELISA Microplates
GREENER_CHOICE
Product Code. 11990657
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Surface Treatment:
Microlon 600 High Binding
Microlon Medium Binding
Non-treated
Unit Size:
Pack of 40
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Surface Treatment unitSize
11990657 Non-sterile Non-treated Pack of 40
10023733 Non-sterile Microlon 600 High Binding Pack of 40
10465385 Sterile Non-treated Pack of 40
13014983 Non-sterile Microlon Medium Binding Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11990657 Supplier Greiner Bio-One Supplier No. 675101
Add to basket
Add to basket
For diagnostics and immunological research applications. Greiner Bio-One 96-Well High Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Crystal clear polystyrene
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Well-to-well spacing: 9mm
  • Specifically tre
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Format Half-Area
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
Quantity 40/Cs.
Well Design Chimney