Learn More
Falcon™ 96-Well Flat Bottom TC-Treated Microplate with Lid
Product Code. 32018025
Change view
Click to view available options
Color:
Black, Clear Bottom
White, Clear Bottom
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
32018025 Black, Clear Bottom Pack of 50
32018024 White, Clear Bottom Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32018025 Supplier Falcon™ Supplier No. 353388
Estimated Shipment: 10-12-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm
  • Opaque walls prevent well-to-well cross-talk
  • Optically clear flat bottom permits direct microscopic viewing
  • Crystal-grade virgin polystyrene
  • Sterilized by gamma irradiation
  • Non-pyrogenic
  • Non-treated polystyrene, standard tissue culture (TC), and Corning™ Primaria™ surface treatments available
  • Convenient, peel-open medical-style packaging for individual product presentation
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
  • Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
TRUSTED_SUSTAINABILITY
Spécification
Barcode No Barcode
Format Standard
Lid Yes
Culture Area 0.32 cm2
Array 8 x 12
Color Black, Clear Bottom
Product Type Microplate
Material Virgin Polystyrene
Volume (Metric) Well 0.37 mL
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Non-sterile
Surface Treatment Tissue Culture
Well Shape Flat Bottom (F-well)
No. per Case 50/Cs.
No. per Pack 10/Pk.
Well Color Clear
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.