missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Conical Bottom (V) Polypropylene Microplates
GREENER_CHOICE

Product Code. p-8015552
Click to view available options
Color:
Black
Red
Yellow
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10582123

Brand: Greiner Bio-One 651209

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Conical Bottom (V) Polypropylene Microplates is free of detectable DNase, RNase, human DNA.

  • Uniform external dimensions
  • Well to well spacing of 9mm
  • Features alphanumeric coding
  • Sealable with adhesive films and heat seal
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens.
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Black
Material Polypropylene
No. of Wells 96
Volume (Metric) Working 50 to 335 μL
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
No. per Pack 10
Quantity 100/Cs.
Temperature Range (Metric) -196°C to +121°C
Well Design Chimney
Dimensions (L x W x H) 128 x 85 x 15 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.