Learn More
Greiner Bio-One 96-well Chimney designed Round (U) Bottom Microplates
Product Code. 10779984
Click to view available options
Color:
Black
Natural
Red
White
Yellow
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Product Code. 10779984
Brand: Greiner Bio-One 650261
Only null left
Add to basket
Add to basket
For high-throughput screening in cell based test systems.
  • Non-pyrogenic
  • Conical well design with round (U) bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Free of detectable DNase, RNase, human DNA
  • Prevents wicking
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Natural
Material Polypropylene
No. of Wells 96
Sterility Sterile
Well Shape Round Bottom (U-well)
Quantity 100/Cs.