Learn More
Greiner Bio-One 96-well Chimney designed Round (U) Bottom Microplates
Product Code. 10440912
Click to view available options
Color:
Black
Natural
Red
White
Yellow
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Product Code. 10440912
Brand: Greiner Bio-One 650209
Only null left
Add to basket
Add to basket
For high-throughput screening in cell based test systems.
  • Non-pyrogenic
  • Conical well design with flat bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Free of detectable DNase, RNase, human DNA
  • Prevents wicking
TRUSTED_SUSTAINABILITY
Specifications
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type Non Binding
Color Black
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 355 μL
Well Shape Round Bottom (U-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
Quantity 100/Cs.
RNase Free Yes
Well Color Black
Well Design Chimney