Learn More
TPP™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
Product Code. 11331714
Change view
Click to view available options
Packaging:
1/Pk., 162/Cs.
6/Pk., 108/Cs.
Unit Size:
Pack of 108
Pack of 162
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
11331714 1/Pk., 162/Cs. Pack of 162
11341774 6/Pk., 108/Cs. Pack of 108
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11331714 Supplier TPP™ Supplier No. 92097
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
In Stock
Add to basket
Add to basket
Color: Clear; Sterility: Sterile; Lid: With Lid
  • Opto-mechanical treatment of growth area for optimal cell adhesion
  • Lid with air-venting system guarantees controlled gas exchange with low evaporation
  • Sloped edge allows placement of lid in one position only
  • Yellow marking area on lid and plate ensure correct lid orientation
  • Ridged grip area ensures secure grasp and prevents from accidentally lifting off lid
  • Clear alpha-numerical labelling between wells for easy orientation during microscopy
  • Growth area on spherical zone only
  • Absolutely flat growth surface
  • Crystal clear transparency
Specifications
Certifications/Compliance ANSI/SLAS 1-2004 through ANSI/SLAS 4-2004
Lid With Lid
Culture Area 0.636 cm2
Packaging 1/Pk., 162/Cs.
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 0.34 mL
Well Shape Round Bottom (U-well)
No. per Case 162, Multi Pack
Dimensions (L x W x H) 128 x 86 x 17 mm