missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ 96 Deep-Well Plate (Autoplate) for MPure-32
Shop All MP Biomedicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
100/Pk.
missing translation for 'unitSize'
Pack of 100
Description
- To use with MPure-32 aNAP system.
- Ideal for 16 or 32 runs.
- Each 96 Deep well plate is designed to receive the sample and reagents for 16 runs.
- Two plates can be installed on the instrument simultaneously.
Specifications
Specifications
| Sample Type | Tissue, Cell Culture, Bacteria |
| Format | 96 Deep-well Plate |
| For Use With (Application) | MPure-32 |
| Quantity | 100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction