Learn More
Applied Biosystems™ 3730xl DNA Analyzer 96-Capillary Array, 50 cm
Shop All Applied Biosystems ProductsDescription
Includes
Each array contains 96 capillaries (50cm in length) for use with the Applied Biosystems 3730XL DNA Analyzer
- Usage per array: 300 runs (with use of Applied Biosystems reagents)
Specifications
Especificaciones
| Capillary Length | 50 cm |
| Content And Storage | Each array contains 96 capillaries (50 cm in length) for use with the Applied Biosystems 3730xl DNA Analyzer. We guarantee the performance of the 3730 Capillary arrays up to 300 runs and within 1 year of purchase. |
| Format | Capillary Array |
| For Use With (Equipment) | 3730xl DNA Analyzer |
| For Use With (Application) | Sequencing |
| Number of Capillaries | 96 Capillaries |
| Product Type | DNA Analyzer 96-Capillary Array |
| Quantity | 1 Array |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
It is possible that the polymer tube fitting may not be screwed into the pump properly, allowing air to get pulled in as the piston retracts. It is also possible that some debris may be stuck in the check valve, locking it in the open position. It may be necessary to detach and carefully attach the polymer tube again and then perform a water wash wizard to remove the bubbles and clear any debris from the check valve.
Leak detected error messages can also occur when the piston on the pump moves further than expected during the movement of the pump. The usual cause for this is a leak on the system, usually due to either an improperly tightened or improperly threaded fitting or a broken capillary. However, if this is not observed, some of the other reasons are:
1.Valve stuck in open position
2.Large air bubble under the piston
3. Polymer:
-Wrong polymer type (e.g., instrument thinks it has POP-7 on it but it really has POP-4).
-Old/degraded polymer
4. Improper/no maintenance on the system
Inspect the system for leaks. If you do not see any leaks on the system, perform a water wash using the water wash wizard and place fresh, non-expired polymer and freshly made 1X Running Buffer on the system. If the problem persists, a service call may be required.
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2.Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3.Using Running Buffer that was:
-Diluted to 1X incorrectly
-Swapped with the water position on the
Autosampler deck
-On the instrument longer than 48 hours
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform a water wash using the water wash wizard and place fresh, non-expired polymer and freshly made 1X Running Buffer on the system. If the problem persists, a service call may be required.
If we do not specify the expiry date or number of uses, the warranty will last for 12 months from the date of shipment.
The arrays are warrantied for a certain number of runs or one year from date of shipment, whichever comes first. If you notice that you are starting to get reduced data quality (e.g., shorter reads, trailing peaks in all 4 colors) even with the Sequencing Standard, these may be signs that the array requires changing.
For Research Use Only. Not for use in diagnostic procedures.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.