missing translation for 'onlineSavingsMsg'
Learn More

Uvex 95281 Low Tune-Up Insole For Uvex 1 and 2 - Width 11

Tune-Up antistatic insole for reducing fatigue and supporting arch

Specifications

Color Black, Blue
For Use With (Equipment) Uvex 1 and 2
Material Microvelour
Disposable Reusable
Gender Unisex
View More Specs

Description

Description

Uvex Tune-Up Comfortable Low Insole 95281

  • Antistatic insole concept for use in safety footwear
  • Designed to reduce fatigue by increasing wearer comfort and enhancing ergonomics by optimum support of arch of feet
  • Good cushioning
  • Relief of pressure marks
  • Non-irritating, comfortable surface structure
  • Breathable and absorbs moisture
  • Antibacterial
  • Antistatic
  • Useable for ESD-footwear
  • Uvex 1, Uvex 2, Uvex 1 Duo, Uvex Xenova™ Light
  • Article No. 95281
  • Standard EN ISO 20345:2011 S1 SRC
  • Sizes: 35 to 52
Specifications

Specifications

Black, Blue
Microvelour
Unisex
Yes
Uvex 1 and 2
Reusable
Standard Width
No
Videos
SDS
Documents

Documents

Product Certifications