missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex 94908 Comfortable Climate Insole For Uvex G2 - Width 11
Comfortable climatic insole with antistatic properties for Uvex 1 G2
POA
Specifications
Color | Black |
---|---|
For Use With (Equipment) | Uvex G2 |
Material | Microvelour |
Disposable | No |
Gender | Unisex |
Description
- Full-surface foot shock absorption
- Highly breathable, moisture-absorbent and quickly re-drying material
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No: 9490.8
- Sizes: 35 to 52
Specifications
Black | |
Microvelour | |
Unisex | |
Yes |
Uvex G2 | |
No | |
Standard width | |
No |