Learn More
uvex 9169 Super F Over-the-Glasses Safety Glasses
Product Code. 15365912 Shop All Uvex Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Lens Tint:
Amber
Clear
Light Brown
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Coating Lens Tint unitSize
15365912 Scratch-resistant Clear Each
17217261 Anti-fog, Scratch-resistant Clear Each
17207261 Anti-fog, Scratch-resistant Light Brown Each
17237261 Scratch-resistant Amber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15365912 Supplier uvex Supplier No. 9169585
Only null left
Add to basket
Edge
Add to basket
These extremely comfortable safety spectacles provide excellent eye area protection, and fit perfectly over conventional prescription lenses. Specs are available in a variety of lens coating and lens tints.
  • Overspecs for users who also wear prescription lenses with an arm-hinge concept
  • Side arms graduate to soft, flexible material for enhanced comfort and grip
  • Unrestricted peripheral vision
Supravision Excellence Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant and chemical-resistant
  • Easy to clean, and less susceptible to soiling
Supravision Sapphire Coating
  • Inside: Scratch-resistant and chemical-resistant
  • Outside: Scratch-resistant and chemical-resistant
  • Non-stick properties allow for easy removal of greasy/watery dirt
Infradur Plus Coating
  • Inside: Anti-fog
  • Outside: Scratch-resistant
  • Minimizes burns due to welding sparks
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 170
Lens Type Supravision Sapphire
Lens Coating Scratch-resistant
Description Shield-lens spectacle
Lens Tint Clear
Product Type W 166 FT CE
Frame Color Black
Model 9169 Super F OTG
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard