Learn More
Hamilton™ 900 Microliter Syringes, RN Termination, No Needle Included
The 900 series syringe is the economical version of the 800 series syringe. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 10 µL. The syringe holder is permanently attached to the syringe barrel. This reduces the initial cost of the syringe but eliminates the possibility of replacement components.
Brand:  Hamilton™ 7648-01
Code : NEW
Additional Details : Weight : 0.05500kg
 View more versions of this product
Product Code. 11590132
  • £147.00 / Each
Estimated Shipment: 31-05-2024
to see stock.
Description
Description
  • Ideal for dispensing volumes from 0.5 µL up to 10 µL
  • Syringe holder is permanently attached to the syringe barrel
Specifications
Specifications
10 μL