missing translation for 'onlineSavingsMsg'
Learn More

9-Vinylcarbazole, 98+%

Product Code. 15482608
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15482608 250 g 250g
15472608 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15482608 Supplier Thermo Scientific Alfa Aesar Supplier No. H56174.30
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1484-13-5 | C14H11N | 193.25 g/mol

9-Vinylcarbazole is used as a monomer in the production of poly(vinylcarbazole), a conductive polymer, in which conductivity is photon-dependent. It is also used in the photoreceptors of photocopiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Vinylcarbazole is used as a monomer in the production of poly(vinylcarbazole), a conductive polymer, in which conductivity is photon-dependent. It is also used in the photoreceptors of photocopiers.

Solubility
Soluble in acetonitrile.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1484-13-5
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00004966,MFCD00134336
InChI Key KKFHAJHLJHVUDM-UHFFFAOYSA-N
Synonym 9-vinylcarbazole, n-vinylcarbazole, 9-vinyl-9h-carbazole, 9h-carbazole, 9-ethenyl, n-vinyl carbazole, vinylcarbazole, 1-vinylcarbazole, poly 9-vinylcarbazole, carbazole, 9-vinyl, poly-n-vinylcarbazole
PubChem CID 15143
IUPAC Name 9-ethenylcarbazole
SMILES C=CN1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 60°C to 65°C
Boiling Point 154°C to 155°C
Quantity 250 g
UN Number UN2811
Beilstein 132988
Solubility Information Soluble in acetonitrile.
Formula Weight 193.24
Percent Purity ≥98%
Chemical Name or Material 9-Vinylcarbazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.