missing translation for 'onlineSavingsMsg'
Learn More

9-Propenyladenine, MedChemExpress

Product Code. 16472768
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16472768

Brand: Medchem Express HY100079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 9-Propenyladenine is a mutagenic impurity in tenofovir disoproxil fumarate. Tenofovir is an antiretroviral drug known as nucleotide analogue reverse transcriptase inhibitors, which block reverse transcriptase, a crucial virus enzyme in HIV-1 and HBV.

9-Propenyladenine is a mutagenic impurity in tenofovir disoproxil fumarate. Tenofovir is an antiretroviral drug known as nucleotide analogue reverse transcriptase inhibitors, which block reverse transcriptase, a crucial virus enzyme in HIV-1 and HBV.

Specifications

Chemical Name or Material 9-Propenyladenine
CAS 1446486-33-4
Color White
Purity Grade Notes Research
Molecular Formula C8H9N5
Quantity 10 mg
Synonym Mutagenic Impurity of Tenofovir Disoproxil Tenofovir Impurity 2
Solubility Information DMSO : ≥ 29 mg/mL (165.53 mM)
SMILES NC1=C2N=CN(/C=C/C)C2=NC=N1
Molecular Weight (g/mol) 175.19
Formula Weight 175.19
Percent Purity 96.69%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.