Learn More
9-Phenyl-2,3,7-trihydroxy-6-fluorone, Thermo Scientific Chemicals
£43.20 - £627.00
Chemical Identifiers
CAS | 975-17-7 |
---|---|
Molecular Formula | C19H12O5 |
Molecular Weight (g/mol) | 320.3 |
MDL Number | MFCD00005048 |
InChI Key | YDCFOUBAMGLLKA-UHFFFAOYSA-N |
Synonym | phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone |
PubChem CID | 70420 |
IUPAC Name | 2,6,7-trihydroxy-9-phenylxanthen-3-one |
SMILES | C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11450403
|
Thermo Scientific Alfa Aesar
A11495.06 |
5 g |
£43.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460403
|
Thermo Scientific Alfa Aesar
A11495.14 |
25 g |
£186.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470403
|
Thermo Scientific Alfa Aesar
A11495.22 |
100 g |
£627.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).
Solubility
Soluble in chloroform.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents. Store at -20°C.
Chemical Identifiers
975-17-7 | |
320.3 | |
YDCFOUBAMGLLKA-UHFFFAOYSA-N | |
70420 | |
C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O |
C19H12O5 | |
MFCD00005048 | |
phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone | |
2,6,7-trihydroxy-9-phenylxanthen-3-one |
Specifications
975-17-7 | |
C19H12O5 | |
40910 | |
Soluble in chloroform. | |
C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O | |
320.3 | |
320.3 |
>300°C | |
MFCD00005048 | |
phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone | |
YDCFOUBAMGLLKA-UHFFFAOYSA-N | |
2,6,7-trihydroxy-9-phenylxanthen-3-one | |
70420 | |
9-Phenyl-2,3,7-trihydroxy-6-fluorone |
Safety and Handling
missing translation for 'einecsNumber' : 213-550-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only