missing translation for 'onlineSavingsMsg'
Learn More

9-Phenanthroic Acid, TRC

Product Code. 30428301
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428301 50 mg -
30421402 10 mg -
30409025 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428301 Supplier Toronto Research Chemicals Supplier No. TRCP33838050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 837-45-6
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.24
Synonym NSC 1922
IUPAC Name phenanthrene-9-carboxylic acid
SMILES OC(=O)c1cc2ccccc2c3ccccc13

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H10O2/c16-15(17)14-9-10-5-1-2-6-11(10)12-7-3-4-8-13(12)14/h1-9H,(H,16,17)
Formula Weight 222.0681
Chemical Name or Material 9-Phenanthroic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.