missing translation for 'onlineSavingsMsg'
Learn More

9-Methylanthracene, 99%, ACROS Organics™

Brand:  Acros Organics 126300100

Code : 29

Additional Details : CAS Number : 779-02-2 Weight : 0.06000kg Transport : UN number : 3077 Chem class : 9 Pack group : III


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10205370

  • £45.10 / 10g


EU Stock 8
Add to basket


Specifications

Chemical Identifiers

9-Methylanthracene
99%
192.26
99%
C15H12

Specifications

9-Methylanthracene
196.0° to 197.0°C (12.0 mmHg)
76.0° to 79.0°C
779-02-2
Glass bottle
Crystals or Crystalline Powder
99%
192.26
1.0660g/mL
Yellow to orange
10g
Authentic
1.066
99%
C15H12
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 9-Methylanthracene

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS