missing translation for 'onlineSavingsMsg'
Learn More

9-Methyl Biotin (mixture of diastereomers), TRC

Product Code. 30481422
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481422 1 mg 1mg
30481244 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481422 Supplier Toronto Research Chemicals Supplier No. TRCM2940751MG
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 415725-35-8
Molecular Formula C11 H18 N2 O3 S
Molecular Weight (g/mol) 258.34
Synonym (3aS,4S,6aR)-Hexahydro-α-methyl-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanoic Acid, 2-Methyl-5-[(3aS,4S,6aR)-2-oxohexahydrothieno[3,4-d]imidazol-4-yl]pentanoic Acid, Biotin Imp. D (EP)
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-2-methylpentanoic acid
SMILES CC(CCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C11H18N2O3S/c1-6(10(14)15)3-2-4-8-9-7(5-17-8)12-11(16)13-9/h6-9H,2-5H2,1H3,(H,14,15)(H2,12,13,16)/t6?,7-,8-,9-/m0/s1
Formula Weight 258.1038
Chemical Name or Material 9-Methyl Biotin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.