missing translation for 'onlineSavingsMsg'
Learn More

9-Hydroxyfluorene, TRC

Product Code. 30136538
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136538 25 g 25g
30009543 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136538 Supplier Toronto Research Chemicals Supplier No. TRCH94249025G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1689-64-1
Molecular Formula C13 H10 O
Molecular Weight (g/mol) 182.22
Synonym Fluorenol
IUPAC Name 9H-fluoren-9-ol
SMILES OC1c2ccccc2c3ccccc13

Specifica

Quantity 25 g
InChI Formula InChI=1S/C13H10O/c14-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)13/h1-8,13-14H
Formula Weight 182.0732
Percent Purity >95%
Chemical Name or Material 9-Hydroxyfluorene
Grade HPLC
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.