missing translation for 'onlineSavingsMsg'
Learn More

9-Hydroxy-pyrido[1,2-a]pyrimidin-4-one (~90%), TRC

Product Code. 30394140
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30394140 10 mg -
30389497 100 mg -
30392449 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394140 Supplier Toronto Research Chemicals Supplier No. TRCH95619810MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123458-49-1
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
IUPAC Name 9-hydroxypyrido[1,2-a]pyrimidin-4-one
SMILES OC1=CC=CN2C(=O)C=CN=C12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H6N2O2/c11-6-2-1-5-10-7(12)3-4-9-8(6)10/h1-5,11H
Formula Weight 162.0429
Chemical Name or Material 9-Hydroxy-pyrido[1,2-a]pyrimidin-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.