missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9-Hydroxy-9-fluorenecarboxylic acid, 97%

Product Code. 11410537
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11410537

Brand: Alfa Aesar™ B22409.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 467-69-6
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00001137
InChI Key GXAMYUGOODKVRM-UHFFFAOYSA-N
Synonym flurenol, 9-hydroxy-9-fluorenecarboxylic acid, flurecol, 9-hydroxy-9h-fluorene-9-carboxylic acid, glycolic acid, diphenylene, 9h-fluorene-9-carboxylic acid, 9-hydroxy, flurecol bsi:iso, unii-4tkt65am8p, 9-hydooxyfluorene-9-carboxylic acid, 9-hydroxy-9-fluorene carboxyli acid
PubChem CID 10087
ChEBI CHEBI:82015
IUPAC Name 9-hydroxyfluorene-9-carboxylic acid
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C(=O)O)O

Specifications

Melting Point ∼164°C (decomposition)
Quantity 100 g
UN Number UN3077
Beilstein 1314711
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 9-Hydroxy-9-fluorenecarboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.