missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethyl pentafluorophenyl carbonate, 98%, Thermo Scientific™

Product Code. 11435976
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435976 25 g 25g
11415976 1 g 1g
11425976 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435976 Supplier Thermo Scientific Alfa Aesar Supplier No. B22297.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88744-04-1
Molecular Formula C21H11F5O3
Molecular Weight (g/mol) 406.308
MDL Number MFCD00013261
InChI Key CBBKZVZOEBSFQX-UHFFFAOYSA-N
Synonym 9-fluorenylmethyl pentafluorophenyl carbonate, fmoc-opfp, 9h-fluoren-9-yl methyl perfluorophenyl carbonate, 9h-fluoren-9-ylmethyl 2,3,4,5,6-pentafluorophenyl carbonate, 9h-fluoren-9-ylmethyl pentafluorophenyl carbonate, carbonic acid 9h-fluoren-9-ylmethyl pentafluorophenyl ester, carbonic acid,9h-fluoren-9-ylmethyl 2,3,4,5,6-pentafluorophenyl ester, acmc-20ecck, fmoc pentafluorophenyl ester, 9-fluorenylmethylpentafluorophenylcarbonate
PubChem CID 2733201
IUPAC Name 9H-fluoren-9-ylmethyl (2,3,4,5,6-pentafluorophenyl) carbonate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)OC4=C(C(=C(C(=C4F)F)F)F)F

Specifications

Melting Point 83°C to 86°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 406.31
Percent Purity 98%
Chemical Name or Material 9-Fluorenylmethyl pentafluorophenyl carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.