missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethyl Chlorocarbonate, TRC

Product Code. 17994365
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994365 10 g 10g
17934055 5 g 5g
17972905 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994365 Supplier Toronto Research Chemicals Supplier No. TRCF47245010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28920-43-6
Molecular Formula C15 H11 Cl O2
Molecular Weight (g/mol) 258.7
Synonym 9H-Fluoren-9-ylmethyl Ester Carbonochloridic Acid, Fluorene-9-methanol, Chloroformate (8CI), (9H-Fluoren-9-ylmethoxy)carbonyl Chloride, 9-Fluorenylmethoxycarbonyl Chloride, 9-Fluorenylmethyl Chlorocarbonate, 9-Fluorenylmethyl Chloroformate, 9H-Fluoren-9-ylmethyl Carbonochloridate, 9H-Fluoren-9-ylmethyl Chloroformate, AminoTag, FMOC Chloride, Fluorenylmethyl Chloroformate, FmocCl
IUPAC Name 9H-fluoren-9-ylmethyl carbonochloridate
SMILES ClC(=O)OCC1c2ccccc2c3ccccc13

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H11ClO2/c16-15(17)18-9-14-12-7-3-1-5-10(12)11-6-2-4-8-13(11)14/h1-8,14H,9H2
Formula Weight 258.0448
Chemical Name or Material 9-Fluorenylmethyl Chlorocarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.