missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethyl 1-benzotriazolyl carbonate, 98%, Thermo Scientific™

Product Code. 11966721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11966721 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11966721 Supplier Thermo Scientific Acros Supplier No. 441510050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 82911-71-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 165°C to 167°C
Color White
Elemental Analysis 68.82 to 72.34% C, 11.46 to 12.05% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C21H15N3O3
Quantity 5 g
Synonym fmoc-obt, 9h-fluoren-9-yl methyl 1h-benzo d 1,2,3 triazol-1-yl carbonate, 9-fluorenylmethyl 1-benzotriazolyl carbonate, benzotriazol-1-yl 9-fluorenylmethyl carbonate, 1,2,3-benzotriazol-1-yl 9h-fluoren-9-ylmethyl carbonate, 1-fmoc-oxy benzotriazole, 1-fmoc-oxy-1h-benzotriazole, 9-fluorenylmethyl benzotriazolycarbonate, 9-fluorenylmethylbenzotriazolylcarbonate, 9-fluorenylmethyl 1-benzotriazolyl &
InChI Key KNSPZTVPSAKHBN-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)ON4C5=CC=CC=C5N=N4
IUPAC Name benzotriazol-1-yl 9H-fluoren-9-ylmethyl carbonate
Molecular Weight (g/mol) 357.37
PubChem CID 5236567
Formula Weight 357.37
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 9-Fluorenylmethyl 1-benzotriazolyl carbonate
Show More Show Less
compliance-icons
Product Identifier
  • 9-Fluorenylmethyl 1-benzotriazolyl carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.