missing translation for 'onlineSavingsMsg'
Learn More

(9-Fluorenyl)triphenylphosphonium bromide, 98%, Thermo Scientific™

Product Code. 11330224
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11330224 5 g 5g
11320224 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11330224 Supplier Thermo Scientific Alfa Aesar Supplier No. L00794.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7253-07-8
MDL Number MFCD00031650
ChEBI CHEBI:40656

Specifications

Melting Point >330°C (decomposition)
Molecular Formula C6H12O6
Quantity 5 g
Beilstein 3897970
Synonym beta-d-allose, beta-d-allopyranose, 3 s-hydroxy-all-cis-8,11,14,17-eicosatetraenoyl-coa, 2r,3r,4r,5s,6r-6-hydroxymethyl oxane-2,3,4,5-tetrol, 1rpj, .beta.-d-allose, d +-allose, b-d-allopyranose, d-allose, beta-d-allose, rare aldohexose sugar
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILES C(C1C(C(C(C(O1)O)O)O)O)O
IUPAC Name (2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular Weight (g/mol) 180.16
PubChem CID 448388
Formula Weight 507.42
Percent Purity 98%
Chemical Name or Material (9-Fluorenyl)triphenylphosphonium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.