missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Fluorenone, 99+%, Thermo Scientific Chemicals
£10.80 - £213.00
Chemical Identifiers
CAS | 486-25-9 |
---|---|
InChI Key | YLQWCDOCJODRMT-UHFFFAOYSA-N |
Synonym | 9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone |
PubChem CID | 10241 |
ChEBI | CHEBI:17922 |
IUPAC Name | fluoren-9-one |
SMILES | C1=CC=C2C(=C1)C3=CC=CC=C3C2=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10589430
|
Thermo Scientific Acros
119200050 |
5 g | Glass Bottle | ||||||
10734041
|
Thermo Scientific Acros
119201000 |
100 g | Plastic bottle |
£21.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10754041
|
Thermo Scientific Acros
119205000 |
500 g | Plastic bottle |
£62.40
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904851
|
Thermo Scientific Acros
119200025 |
2.5 kg | Plastic bottle |
£213.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
486-25-9 | |
9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone | |
CHEBI:17922 | |
C1=CC=C2C(=C1)C3=CC=CC=C3C2=O |
YLQWCDOCJODRMT-UHFFFAOYSA-N | |
10241 | |
fluoren-9-one |
Specifications
486-25-9 | |
Yellow | |
99+% | |
9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone | |
C1=CC=C2C(=C1)C3=CC=CC=C3C2=O | |
180.206 | |
CHEBI:17922 | |
≥99.0% | |
9-Fluorenone |
81°C to 85°C | |
Authentic | |
C13H8O | |
YLQWCDOCJODRMT-UHFFFAOYSA-N | |
fluoren-9-one | |
10241 | |
180.21 | |
Crystalline Powder or Flakes |
RUO – Research Use Only