missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenol, 98+%

Product Code. 15468799
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468799 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15468799 Supplier Thermo Scientific Alfa Aesar Supplier No. L01917.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1689-64-1 | C13H10O | 182.222 g/mol

9-Fluorenol is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenol is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Insoluble in water.

Notes
Protect from moisture, heat. Store in cool place. Keep container tightly
TRUSTED_SUSTAINABILITY

Specifications

CAS 1689-64-1
Melting Point 155°C to 157°C
Molecular Formula C13H10O
MDL Number MFCD00001135
Quantity 25 g
Beilstein 1869799
Synonym 9-hydroxyfluorene, 9-fluorenol, fluoren-9-ol, fluorenol, 9-hydroxy fluorene, diphenylene carbinol, unii-bv0q72r613, hydroxyfluoren, 9-hydroxy-fluoren, 9h-fluorene-9-ol
Solubility Information Insoluble in water.
InChI Key AFMVESZOYKHDBJ-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)O
IUPAC Name 9H-fluoren-9-ol
Molecular Weight (g/mol) 182.222
PubChem CID 74318
ChEBI CHEBI:16904
Formula Weight 182.22
Percent Purity ≥98%
Chemical Name or Material 9-Fluorenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.