missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenemethanol, TRC

Product Code. 30393913
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393913 50 g
30397596 100 g
30399820 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393913 Supplier Toronto Research Chemicals Supplier No. TRCF53243550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24324-17-2
Molecular Formula C14 H12 O
Molecular Weight (g/mol) 196.24
Synonym FMOC-hydroxy
IUPAC Name 9H-fluoren-9-ylmethanol
SMILES OCC1c2ccccc2c3ccccc13

Specifications

Quantity 50 g
InChI Formula InChI=1S/C14H12O/c15-9-14-12-7-3-1-5-10(12)11-6-2-4-8-13(11)14/h1-8,14-15H,9H2
Formula Weight 196.0888
Chemical Name or Material FMOC-hydroxy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.