missing translation for 'onlineSavingsMsg'
Learn More

9-Ethynyl-9-fluorenol, 98%

Product Code. 11322237
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11322237

Brand: Thermo Scientific Alfa Aesar L09717.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13461-74-0 | C15H10O | 206.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13461-74-0
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.244
MDL Number MFCD00041555
InChI Key MMZVVJGCZZAWBN-UHFFFAOYSA-N
Synonym 9-ethynyl-9-fluorenol, 9-ethynyl-9h-fluoren-9-ol, acmc-209bwd, 9-ethynyl-9-hydroxyfluorene, 9-ethinyl-9-hydroxy-fluorene, 9h-fluoren-9-ol,9-ethynyl, 9-ethynyl-9h-fluoren-9-ol #, mmzvvjgczzawbn-uhfffaoysa, 9-hydroxy-9-fluorenyl acetylene, 9-ethynyl-9-fluorenol,
PubChem CID 518771
IUPAC Name 9-ethynylfluoren-9-ol
SMILES C#CC1(C2=CC=CC=C2C3=CC=CC=C31)O

Specifications

Melting Point 109°C to 110°C
Quantity 5 g
Beilstein 2104685
Formula Weight 206.25
Percent Purity 98%
Chemical Name or Material 9-Ethynyl-9-fluorenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.