missing translation for 'onlineSavingsMsg'
Learn More

9-Dihydro-13-acetylbaccatin III, MedChemExpress

Product Code. 16477956
Encompass_Preferred
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
This item is not returnable. View return policy

Product Code. 16477956

Brand: Medchem Express HY77434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 9-Dihydro-13-acetylbaccatin III (9-DHAB III) is an intermediate for taxol analog preparations.

9-Dihydro-13-acetylbaccatin III (9-DHAB III) is an intermediate for taxol analog preparations.

Specifications

Chemical Name or Material 9-Dihydro-13-acetylbaccatin III
CAS 142203-65-4
Color White
Purity Grade Notes Research
Molecular Formula C33H42O12
Quantity 100 mg
Synonym 9-DHAB III 13-Acetyl-9-dihydrobaccatin III
Solubility Information DMSO : 20 mg/mL (31.71 mM; Need ultrasonic)
SMILES O[C@@H](C1)[C@]2(C)[C@]([C@@]3(OC(C)=O)[C@]1([H])OC3)([H])[C@H](OC(C4=CC=CC=C4)=O)[C@]5(O)C[C@H](OC(C)=O)C(C)=C(C(C)5C)[C@@H](OC(C)=O)[C@@H]2O
Molecular Weight (g/mol) 630.68
Formula Weight 630.68
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.