missing translation for 'onlineSavingsMsg'
Learn More

9-Decen-1-ol, TRC

Product Code. 30367804
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367804 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367804 Supplier Toronto Research Chemicals Supplier No. TRCD22559525G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-Decen-1-ol
CAS 13019-22-2
Molecular Formula C10H20O
Quantity 25 g
Synonym 9-Decenol, Decylenic Alcohol, NSC 103158, Rosalva, Trepanol, ω-Decen-1-ol
InChI Formula InChI=1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h2,11H,1,3-10H2
SMILES OCCCCCCCCC=C
IUPAC Name dec-9-en-1-ol
Molecular Weight (g/mol) 156.27
Formula Weight 156.1514
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.