missing translation for 'onlineSavingsMsg'
Learn More

9-Decen-1-ol, 96%

Product Code. 15254644 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15254644 100 g Glass bottle 100g
15214644 5 g Glass bottle 5g
15234644 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15254644 Supplier Thermo Scientific Acros Supplier No. 152641000
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 13019-22-2 | C10H20O | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13019-22-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
InChI Key QGFSQVPRCWJZQK-UHFFFAOYSA-N
Synonym 9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol
PubChem CID 25612
IUPAC Name dec-9-en-1-ol
SMILES C=CCCCCCCCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -13.0°C
Color Orange-Red to Red
Density 0.8450g/mL
Boiling Point 234.0°C to 238.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (main isomer) (GC)
Packaging Glass bottle
Linear Formula H2C=CH(CH2)8OH
Refractive Index 1.4470 to 1.4490
Quantity 100 g
Beilstein 01,451
Specific Gravity 0.845
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Viscosity 11.4 mPa.s (20°C)
Formula Weight 156.27
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 9-Decen-1-ol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 9-Decen-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.