missing translation for 'onlineSavingsMsg'
Learn More

9-Decen-1-ol, 90+%

Product Code. 11424067
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424067 25 g 25g
11434067 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424067 Supplier Thermo Scientific Alfa Aesar Supplier No. A19490.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13019-22-2 | C10H20O | 156.269 g/mol

It finds it uses in a wide range of fragrances, especially in fine fragrances and soaps, in floral, rose petal or herbal fragrances. 9-Decen-1-ol is used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semi fluorinated isoprene) block copolymers with -CF2H-terminated side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it uses in a wide range of fragrances, especially in fine fragrances and soaps, in floral, rose petal or herbal fragrances. 9-Decen-1-ol is used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semi fluorinated isoprene) block copolymers with -CF2H-terminated side groups.

Solubility
Soluble in water at 20°C 0.16g/L. Soluble in alcohol, dipropylene glycol, paraffin oil.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13019-22-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00002992
InChI Key QGFSQVPRCWJZQK-UHFFFAOYSA-N
Synonym 9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol
PubChem CID 25612
IUPAC Name dec-9-en-1-ol
SMILES C=CCCCCCCCCO

Specifications

Melting Point -13°C
Density 0.847
Boiling Point 235°C
Flash Point 104°C (219°F)
Odor Pleasant
Refractive Index 1.448
Quantity 25 g
Beilstein 1750928
Solubility Information Soluble in water at 20°C 0.16g/L. Soluble in alcohol,dipropylene glycol,paraffin oil.
Formula Weight 156.27
Percent Purity ≥90%
Chemical Name or Material 9-Decen-1-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.