missing translation for 'onlineSavingsMsg'
Learn More

9-cis-Retinoic acid

Product Code. 15474629
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474629 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15474629 Supplier Thermo Scientific Alfa Aesar Supplier No. J62219.#0
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5300-03-8 | C20H28O2 | 300.44 g/mol

9-cis-Retinoic acid is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. 9-cis-Retinoic acid is an inhibitor of Cox-2 and an activator of RAR α, RAR β, RAR γ and RXR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-cis-Retinoic acid is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. 9-cis-Retinoic acid is an inhibitor of Cox-2 and an activator of RAR α, RAR β, RAR γ and RXR.

Solu
TRUSTED_SUSTAINABILITY

Specifications

CAS 5300-03-8
Melting Point 178°C to 180°C
Molecular Formula C20H28O2
MDL Number MFCD00270072
Quantity 5 mg
Sensitivity Light and moisture sensitive
Merck Index 14,249
Synonym alitretinoin, 9-cis-retinoic acid, panretin, 9-cis-tretinoin, panrexin, panretyn, panretin gel, toctino, 9-cis retinoic acid, 9 z-retinoic acid
Solubility Information Soluble in chloroform (50mg/ml),DMSO (∽25mg/ml),ethanol (25mg/ml),and acetone (25mg/ml). Insoluble in water.
InChI Key SHGAZHPCJJPHSC-YCNIQYBTSA-N
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
IUPAC Name (2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
Molecular Weight (g/mol) 300.44
PubChem CID 449171
ChEBI CHEBI:50648
Formula Weight 300.44
Physical Form Powder
Chemical Name or Material 9-cis-Retinoic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 9-cis-Retinoic acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 9-cis-Retinoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.