missing translation for 'onlineSavingsMsg'
Learn More

9-Carboxymethoxymethylguanine, TRC

Product Code. 17973415
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17973415 5 mg 5mg
17921025 25 mg 25mg
17924375 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973415 Supplier Toronto Research Chemicals Supplier No. TRCC1799905MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80685-22-9
Molecular Formula C8 H9 N5 O4
Molecular Weight (g/mol) 239.19
Synonym Acetic acid, 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-, Acetic acid, [(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]- (9CI), 9-Carboxymethoxymethylguanine
IUPAC Name 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]acetic acid
SMILES NC1=Nc2c(ncn2COCC(=O)O)C(=O)N1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C8H9N5O4/c9-8-11-6-5(7(16)12-8)10-2-13(6)3-17-1-4(14)15/h2H,1,3H2,(H,14,15)(H3,9,11,12,16)
Formula Weight 239.0655
Chemical Name or Material 9-Carboxymethoxymethylguanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.