missing translation for 'onlineSavingsMsg'
Learn More

9-Bromofluorene, 97+%

Product Code. 11334629
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334629 5 g 5g
11478633 50 g 50g
11488633 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334629 Supplier Thermo Scientific Alfa Aesar Supplier No. A10428.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1940-57-4 | C13H9Br | 245.119 g/mol

9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Air sensitive. Stable under normal conditions. Preserve in light-resistant and well-closed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1940-57-4
Molecular Formula C13H9Br
Molecular Weight (g/mol) 245.119
MDL Number MFCD00001133
InChI Key AHCDKANCCBEQJJ-UHFFFAOYSA-N
Synonym 9-bromofluorene, fluorene, 9-bromo, 9h-fluorene, 9-bromo, 9-fluorenyl bromide, monobromofluorene, 9-bromo-fluoren, 9-brom-9h-fluoren, acmc-1bqkc, 9-bromanyl-9h-fluorene
PubChem CID 16024
IUPAC Name 9-bromo-9H-fluorene
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)Br

Specifications

Melting Point 101°C to 105°C
Odor Irritating
Quantity 5 g
UN Number UN3261
Beilstein 2047220
Solubility Information Insoluble in water.
Formula Weight 245.12
Percent Purity ≥98%
Chemical Name or Material 9-Bromofluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.