missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-9-phenylfluorene, 97%, Thermo Scientific Chemicals

Catalog Number p-4089986
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
55135-66-5
C19H13Br
321.21
MFCD00075522
HYQXNCDBSALQLB-UHFFFAOYSA-N
9-bromo-9-phenyl-9h-fluorene, 9-brom-9-phenyl-9h-fluoren, zlchem 855, acmc-209ll7, 9-bromo-9-phenyl-fluorene, 9-bromanyl-9-phenyl-fluorene, hyqxncdbsalqlb-uhfffaoysa, 9-bromo-9-phenylfluorene, 9h-fluorene, 9-bromo-9-phenyl
231624
9-bromo-9-phenylfluorene
C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)Br
This item is not returnable. View return policy
55135-66-5
C19H13Br
321.21
MFCD00075522
HYQXNCDBSALQLB-UHFFFAOYSA-N
9-bromo-9-phenyl-9h-fluorene, 9-brom-9-phenyl-9h-fluoren, zlchem 855, acmc-209ll7, 9-bromo-9-phenyl-fluorene, 9-bromanyl-9-phenyl-fluorene, hyqxncdbsalqlb-uhfffaoysa, 9-bromo-9-phenylfluorene, 9h-fluorene, 9-bromo-9-phenyl
231624
9-bromo-9-phenylfluorene
C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 99°C to 101°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 321.21
Percent Purity 97%
Chemical Name or Material 9-Bromo-9-phenylfluorene
compliance-icons
Product Identifier
  • 9-Bromo-9-phenylfluorene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'