missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-9-phenylfluorene, 96%

Product Code. 11379055
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379055 1 g 1g
11389055 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379055 Supplier Thermo Scientific Alfa Aesar Supplier No. L14225.03
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 55135-66-5 | C19H13Br | 321.217 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55135-66-5
Molecular Formula C19H13Br
Molecular Weight (g/mol) 321.217
MDL Number MFCD00075522
InChI Key HYQXNCDBSALQLB-UHFFFAOYSA-N
Synonym 9-bromo-9-phenyl-9h-fluorene, 9-brom-9-phenyl-9h-fluoren, zlchem 855, acmc-209ll7, 9-bromo-9-phenyl-fluorene, 9-bromanyl-9-phenyl-fluorene, hyqxncdbsalqlb-uhfffaoysa, 9-bromo-9-phenylfluorene, 9h-fluorene, 9-bromo-9-phenyl
PubChem CID 231624
IUPAC Name 9-bromo-9-phenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)Br

Specifications

Melting Point 94°C to 99°C
Quantity 1 g
UN Number UN3261
Beilstein 2561445
Formula Weight 321.23
Percent Purity 96%
Chemical Name or Material 9-Bromo-9-phenylfluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.