missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-1-nonanol, 98%

Product Code. 15497918
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497918 5 g 5g
15407928 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497918 Supplier Thermo Scientific Alfa Aesar Supplier No. H61393.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 55362-80-6 | C9H19BrO | 223.15 g/mol

9-Bromo-1-nonanol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Bromo-1-nonanol is used as a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane, ethyl acetate and hexane.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55362-80-6
Molecular Formula C9H19BrO
Molecular Weight (g/mol) 223.15
MDL Number MFCD00063348
InChI Key USJDOLXCPFASNV-UHFFFAOYSA-N
Synonym 9-bromo-1-nonanol, 1-nonanol, 9-bromo, 9-bromo-nonan-1-ol, nonamethylene bromohydrin, 9-bromnonan-1-ol, 9-bromononanol, 9-bromononyl alcohol, pubchem23429, 9-bromo nonanol, acmc-1axfq
PubChem CID 108700
IUPAC Name 9-bromononan-1-ol
SMILES OCCCCCCCCCBr

Specifications

Melting Point 33°C to 35°C
Boiling Point 125°C to 126°C (2 mmHg)
Flash Point 109°C (228°F)
Linear Formula Br(CH2)9OH
Quantity 5 g
Beilstein 1737525
Solubility Information Soluble in dichloromethane,ethyl acetate and hexane.
Formula Weight 223.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 9-Bromo-1-nonanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.