missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosyladenine-5'-monophosphate, 99%, Thermo Scientific Chemicals

Product Code. p-7043126
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15410467

Brand: Thermo Scientific Alfa Aesar J65653.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Disrupts viral DNA synthesis. Competitive viral DNA polymerase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disrupts viral DNA synthesis. Competitive viral DNA polymerase inhibitor.

Solubility
Soluble in water at 4mg/ml

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29984-33-6
Molecular Formula C10H14N5O7P
Molecular Weight (g/mol) 347.22
MDL Number MFCD00069723
InChI Key UDMBCSSLTHHNCD-UHTZMRCNSA-N
Synonym vidarabine monophosphate, ara-amp, vidarabine phosphate, adenine arabinoside monophosphate, 9-5-o-phosphono-beta-d-arabinofuranosyl adenine, arabinofuranosyladenine monophosphate, vidarabine 5'-monophosphate, vidarabine-5'-monophosphate, vidarabine phosphate usan, arabinosyladenine monophosphate
PubChem CID 34768
IUPAC Name [(2R,3S,4S,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@@H]3O)C2=NC=N1

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water at 4mg/ml
Formula Weight 347.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 9-beta-D-Arabinofuranosyladenine-5'-monophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.