missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosyladenine-5'-monophosphate, 99%, Thermo Scientific Chemicals

Product Code. 15410467
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410467 1 g 1g
15420467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410467 Supplier Thermo Scientific Alfa Aesar Supplier No. J65653.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Disrupts viral DNA synthesis. Competitive viral DNA polymerase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disrupts viral DNA synthesis. Competitive viral DNA polymerase inhibitor.

Solubility
Soluble in water at 4mg/ml

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29984-33-6
Molecular Formula C10H14N5O7P
Molecular Weight (g/mol) 347.22
MDL Number MFCD00069723
InChI Key UDMBCSSLTHHNCD-UHTZMRCNSA-N
Synonym vidarabine monophosphate, ara-amp, vidarabine phosphate, adenine arabinoside monophosphate, 9-5-o-phosphono-beta-d-arabinofuranosyl adenine, arabinofuranosyladenine monophosphate, vidarabine 5'-monophosphate, vidarabine-5'-monophosphate, vidarabine phosphate usan, arabinosyladenine monophosphate
PubChem CID 34768
IUPAC Name [(2R,3S,4S,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@@H]3O)C2=NC=N1

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water at 4mg/ml
Formula Weight 347.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 9-beta-D-Arabinofuranosyladenine-5'-monophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.