missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosyl-2-fluoroadenine-5'-monophosphate, 99%, Thermo Scientific™

Product Code. 15450417
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450417 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15450417 Supplier Thermo Scientific Alfa Aesar Supplier No. J65154.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in DMSO or water at 5mg/ml

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75607-67-9
Molecular Formula C10H13FN5O7P
Molecular Weight (g/mol) 365.21
MDL Number MFCD00866418
InChI Key GIUYCYHIANZCFB-FJFJXFQQSA-N
Synonym fludarabine phosphate, fludara, fludarabine 5'-monophosphate, fludura, oforta, 2-fluoro-ara amp, fludarabine monophosphate, famp, unii-1x9vk9o1sc
PubChem CID 30751
ChEBI CHEBI:63599
SMILES NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@@H]3O)C2=NC(F)=N1

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in DMSO or water at 5mg/ml
IUPAC Name [(2R,3S,4S,5R)-5-(6-amino-2-fluoropurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
Formula Weight 365.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 9-beta-D-Arabinofuranosyl-2-fluoroadenine-5'-monophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.