missing translation for 'onlineSavingsMsg'
Learn More

9-Benzhydrylidene-10-anthrone, 95%, Thermo Scientific™

Product Code. 10650155 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10650155 1 g 1g
10220403 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10650155 Supplier Thermo Scientific Acros Supplier No. 388260010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 667-91-4
Molecular Formula C27H18O
Molecular Weight (g/mol) 358.44
MDL Number MFCD00019172
InChI Key SPOJSTUMWGMCFP-UHFFFAOYSA-N
PubChem CID 69586
IUPAC Name 10-benzhydrylideneanthracen-9-one
SMILES C1=CC=C(C=C1)C(=C2C3=CC=CC=C3C(=O)C4=CC=CC=C42)C5=CC=CC=C5

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 203.0°C to 207.0°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 07, 551
Synonym 9-benzhydrylidene-10-anthrone, 10-diphenylmethylene anthracen-9 10h-one, 10-diphenylmethylidene anthracen-9-one, anthrafuchson, 10-benzhydrylidene-9-anthrone, 10-diphenylmethylene anthracen-9-one, 9-diphenylmethylene-10 9h-anthracenone, 9 10h-anthracenone,10-diphenylmethylene, 10-diphenylmethylene-9 10h-anthracenone #, 9 10h-anthracenone, 10-diphenylmethylene
Formula Weight 358.44
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzhydrylidene-10-anthrone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.