missing translation for 'onlineSavingsMsg'
Learn More

9-Anthracenepropanoic Acid Ethyl Ester, TRC

Product Code. 30339634
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30339634 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339634 Supplier Toronto Research Chemicals Supplier No. TRCA678440100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-Anthracenepropanoic Acid Ethyl Ester
CAS 109690-74-6
Molecular Formula C24 H20 O
Quantity 100 mg
Synonym 3-(9-Anthracenyl)propionic Acid Ethyl Ester, 3-(9-Anthryl)propionic Acid Ethyl Ester
InChI Formula InChI=1S/C24H20O/c1-17-10-12-18(13-11-17)24(25)15-14-23-21-8-4-2-6-19(21)16-20-7-3-5-9-22(20)23/h2-13,16H,14-15H2,1H3
SMILES Cc1ccc(cc1)C(=O)CCc2c3ccccc3cc4ccccc24
IUPAC Name 3-anthracen-9-yl-1-(4-methylphenyl)propan-1-one
Molecular Weight (g/mol) 324.415
Formula Weight 324.151
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.