missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Amino-1,2,3,4-tetrahydroacridin-1-ol Maleate, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 9-Amino-1,2,3,4-tetrahydro-acridin-1-ol Maleate |
| CAS | 118909-22-1 |
| Molecular Formula | C13 H14 N2 O . C4 H4 O4 |
| Quantity | 500 mg |
| Synonym | 1-Acridinol, 9-amino-1,2,3,4-tetrahydro-, (2Z)-2-butenedioate (1:1) (ACI), 1-Acridinol, 9-amino-1,2,3,4-tetrahydro-, (2Z)-2-butenedioate (1:1) (salt) (9CI), 1-Acridinol, 9-amino-1,2,3,4-tetrahydro-, (±)-, (Z)-2-butenedioate (1:1) (salt) (ZCI), 1-Acridinol, 9-amino-1,2,3,4-tetrahydro-, (Z)-2-butenedioate (1:1) (salt), HP 029, Mentane, P 83-6029A, Velnacrine hydrogen maleate, Velnacrine maleate, 1-Hydroxytacrine maleate, 9-Amino-1,2,3,4-tetrahydroacridin-1-ol maleate |
| InChI Formula | InChI=1S/C13H14N2O.C4H4O4/c14-13-8-4-1-2-5-9(8)15-10-6-3-7-11(16)12(10)13;5-3(6)1-2-4(7)8/h1-2,4-5,11,16H,3,6-7H2,(H2,14,15);1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | Nc1c2C(O)CCCc2nc3ccccc13.OC(=O)\C=C/C(=O)O |
| IUPAC Name | 9-amino-1,2,3,4-tetrahydroacridin-1-ol;(Z)-but-2-enedioic acid |
| Molecular Weight (g/mol) | 330.34 |
| Formula Weight | 330.1216 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction